Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant Arginase 1
  89144-056
 :  
Human Recombinant Arginase 1
  89144-056
 :  201-081-C020
 :  

 

  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Size:
    20 µg
  • Form:
    Liquid
  • Enzyme name:
    Arginase 1
  • Enzyme activity:
    ~2.0 U/µg protein. One unit is defined as the amount of enzyme that converts 1 µmol of L-arginine to L-ornithine and urea per min. at 37 °C, pH 9.5 (R.T. Schminke, et al.; J. Biol. Chem. 238, 1012 (1962)).
  • Purity:
    ≥90% (SDS-PAGE)
  • Formulation:
    In 10 mM TRIS-HCl, pH 7.5, containing 1 mM β-mercaptoethanol, 1 mM MnCl₂ and 50% glycerol.
  • Cat. no.:
    89144-056
  • Supplier No.:
    201-081-C020

 

 

Produced in E. coli. Full length human arginase I.