Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Caspase-2 (from E. coli)
Human Recombinant Caspase-2 (from E. coli)
Supplier:  Enzo Life Sciences
Certificates
undefined
Human Recombinant Caspase-2 (from E. coli)
Supplier:  Enzo Life Sciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Form:
    Lyophilized
  • Reconstitution instructions:
    Reconstitute to 1 U/µl with PBS containing 15% glycerol.
  • Enzyme name:
    Caspase-2
  • Enzyme activity:
    10000 - 13000 U/mg protein. One unit is defined as the amount of enzyme that cleaves 1 nmol of the caspase substrate VDVAD-pNA per hour at 37 °C in reaction solution containing 50 mM HEPES, pH 7.2, 50 mM sodium chloride, 0.1% CHAPS, 10 mM EDTA, 5% glycerol and 10 mM DTT.
  • Purity:
    ≥95% (SDS-PAGE)

Specifications

Frequently Bought Together