Human MMP2 proenzyme (from Fibroblasts)
Catalog #: 89143-928
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Source:Fibroblasts
- Species:Human
- Size:5 µg
- Biological activity:≥850mU/mg protein (Y. Masui, et al.; Biochem. Med. 17, 215 (1977)). One unit is defined as the amount of enzyme that hydrolyzes 1µmol Dnp-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg-OH per min. at 37°C, pH 7.0.
- Protein/peptide name:MMP2 proenzyme
- Purity:>90% (SDS-PAGE, Western blot)
- Formulation:Liquid. In 50mM TRIS-HCl, pH 7.0, containing 200mM NaCl, 5mM CaCl2, 1µM ZnCl2, 0.05% BRIJ 35 and 0.05% sodium azide.
- Purification:No other MMP contaminants are detectable.
- Cat. no.:89143-928
- Supplier No.:200-419-C005
Specifications
About this item
Isolated from human rheumatoid synovial fibroblasts. Requires activation.
Customers Who Bought This Also Bought