To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Native
- Source:Plasma
- Species:Human
- Size:1 mg
- Reconstitution instructions:Reconstitute with distilled water.
- Protein/peptide name:Ceruloplasmin
- Purity:>95% (SDS-PAGE)
- Formulation:Lyophilized from 50mM potassium phosphate, pH 6.8, containing 100mM potassium chloride, 20mM ε-aminocaproic acid and 5mM EDTA.
- Purification:Plasma has been tested negative for HBsAg and for antibodies to HIV and HCV.
- Cat. no.:89143-920
- Supplier No.:200-089-M001
Specifications
About this item
Serum copper transport protein
Isolated from human plasma.
Serum copper transport and iron-oxidizing protein. Expressed in plasma at concentrations of 1-5µM. Contains six copper centers and is known to oxidize amines in a process coupled to the reduction of molecular oxygen. Has ferroxidase activity that is responsible for the oxidation of ferrous iron to its ferric form, which is necessary for efficient iron efflux from the cell (e.g. during hypoxia). Has previously been considered a target for nitric oxide (NO) and an inhibitor of endothelial nitric oxide synthase (eNOS/NOS III). Catalyzes S-nitrosothiol formation in cell culture media. Is a NO oxidase and nitrite synthase that determines endocrine NO homeostasis.