Nociceptin
Supplier: Enzo Life Sciences
Certificates
About this item
ORL1 receptor agonist
ORL1 receptor agonist. The ORL1 receptor structurally resembles opioid receptors and is negatively coupled with adenylyl cyclase.
Ordering information: Quantity: *Sold as net peptide weight
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.