Order Entry
Canada
Orders LinkContactUsLinkComponent
Mouse Recombinant HSP60 (from <i>E. coli</i>)

Mouse Recombinant HSP60 (from E. coli)

Supplier:   Enzo Life Sciences
Mouse Recombinant HSP60 (from <i>E. coli</i>)
Supplier:  Enzo Life Sciences
Certificates
Mouse Recombinant HSP60 (from <i>E. coli</i>)

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Produced in E. coli.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 89141-900
  • 89141-902
  • Protein/peptide name:
  • HSP60
  • HSP60
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • Endotoxin level:
  • Low
  • Low
  • Endotoxin content:
  • <50EU/mg purified protein (LAL test)
  • Purity:
  • >90% (SDS-PAGE; Western blot)
  • Form:
  • Liquid
  • Formulation:
  • In 10mM TRIS, pH 7.5, containing 150mM sodium chloride, 1mM EDTA, and 1mM DTT.
  • Purification:
  • Purified by multi-step chromatography.

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 50 µg
    Log in for availability
    Log in for availability
    Each
    Each - $761.55
    $761.55
    Each
       $761.55
    Minimum order quantity is 1

    Supplier Number: ADI-ESP-741-D
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    89141-900
    Protein/peptide name
    HSP60
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Endotoxin level
    Low
    Endotoxin content
    <50EU/mg purified protein (LAL test)
    Purity
    >90% (SDS-PAGE; Western blot)
    Form
    Liquid
    Formulation
    In 10mM TRIS, pH 7.5, containing 150mM sodium chloride, 1mM EDTA, and 1mM DTT.
    Purification
    Purified by multi-step chromatography.
  • 200 µg
    Log in for availability
    Log in for availability
    Each
    Each - $1,775.91
    $1,775.91
    Each
       $1,775.91
    Minimum order quantity is 1

    Supplier Number: ADI-ESP-741-F
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    89141-902
    Protein/peptide name
    HSP60
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Source
    E. coli
    Endotoxin level
    Low
    Endotoxin content
    Purity
    Form
    Formulation
    Purification
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.