Human Recombinant HSP60 (from E. coli)
Supplier: Enzo Life Sciences
About this item
Produced in E. coli.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Produced in E. coli.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 89141-884
- 89141-886
- Protein/peptide name:
- HSP60
- HSP60
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Endotoxin level:
- Low
- Low
- Endotoxin content:
- <50EU/mg purified protein (LAL test)
- Purity:
- >90% (SDS-PAGE; Western blot)
- Form:
- Liquid
- Formulation:
- In Dulbecco’s PBS.
- Purification:
- Purified by multi-step chromatography.
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: ADI-ESP-540-DItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89141-884Protein/peptide nameHSP60Protein/peptide typeRecombinantSpeciesHumanSourceE. coliEndotoxin levelLowEndotoxin content<50EU/mg purified protein (LAL test)Purity>90% (SDS-PAGE; Western blot)FormLiquidFormulationIn Dulbecco’s PBS.PurificationPurified by multi-step chromatography. - Supplier Number: ADI-ESP-540-FItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.89141-886Protein/peptide nameHSP60Protein/peptide typeRecombinantSpeciesHumanSourceE. coliEndotoxin levelLowEndotoxin contentPurityFormFormulationPurification
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



