PKA kinase activity kit, Enzo Life Sciences
Catalog # 89141-834
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:96 wells
- Cat. no.:89141-834
- Supplier No.:EKS-390A
Specifications
About this item
For the quantitative determination of PKA kinase activity in partially purified, purified, or crude enzyme preparations from any species.
The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2