Order Entry
Canada
ContactUsLinkComponent
Hydrogen Peroxide Fluorometric Detection Kit, Enzo Life Sciences
Hydrogen Peroxide Fluorometric Detection Kit, Enzo Life Sciences
Catalog # 89141-682
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Hydrogen Peroxide Fluorometric Detection Kit, Enzo Life Sciences
Catalog # 89141-682
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-907-028
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Hydrogen peroxide fluorometric detection kit
  • Size:
    500 tests
  • Storage conditions:
    +4 °C (long term)
  • Species reactivity:
    Species independent
  • Sensitivity:
    0 - 10 µM (H2O2); 0 - 25 mU/ml (peroxidase)
  • Cat. no.:
    89141-682
  • Application:
    Fluorescent detection, HTS
  • Supplier No.:
    907-028

Specifications

About this item

The Hydrogen Peroxide fluorometric detection kit provides fast results in just 10 minutes for the measurement of either hydrogen peroxide or peroxidase activity in a convenient one-step homogeneous assay.

  • Flexible in measure hydrogen peroxide or peroxidase activity
  • Higher throughput analysis
  • Fast results

For the measurement of hydrogen peroxide in cell lysates and culture supernatants from any species.

Delivery: Reaction buffer concentrate, Detection Reagent, Hydrogen peroxide, Horseradish peroxidase