Benchtop Conductivity/TDS/Salinity Meter, Sper Scientific
97039-650
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Benchtop Conductivity/TDS/Salinity Meter
- Environmentally Preferable:
- Dimensions:217 × 168 × 58 mm (8.5 × 6.5 × 2.25")
- Weight:533 g (18 oz.)
- Cat. no.:97039-650
- Supplier No.:860032
Rugged housing with large LCD displaying the parameter being measured together with time, date and temperature (in °C or °F).
- Automatic or manual temperature compensation
- Automatic or manual ranging, automatic buffer recognition
- Digital and analog outputs
- Min/max and 99 data points
- Data hold
- 5 calibration points
- Electrode status indication
Operates on AC voltage (no batteries).
: 5-year warranty. Comes ready to use with ATC conductivity electrode, electrode holder arm, AC adaptor, computer cable and software.