ContactUsLinkComponent
Advanced pH Meter Kit, Sper Scientific
Advanced pH Meter Kit, Sper Scientific
Catalog # 97036-028
Supplier:  Sper Scientific
undefined
Advanced pH Meter Kit, Sper Scientific
Catalog # 97036-028
Supplier:  Sper Scientific
Supplier Number:  850056
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Advanced pH Meter Kit
  • Environmentally Preferable:
  • Cat. no.:
    97036-028
  • Supplier No.:
    850056
Specifications
About this item
Advanced pH Meter model 850055 reads either pH (in 0.01 resolution) or mV in large digits, with time, date, and temperature (in °C or °F) shown simultaneously on the lower display
Also indicates stable reading, low battery and displays calibration data. Scroll 99 data points with time stamps and min/max readings directly on the backlit LCD. Features hold, up to 5 point calibration, automatic buffer recognition, manual or automatic temperature compensation, and a USB port.
Ordering information: 5-year warranty. Kit includes Advanced pH Meter model 850055 in a hard carrying case complete with a detachable temperature compensated pH probe with 3½’ cable, pH buffers 4,7,10 and distilled water.