Order Entry
Canada
Orders LinkContactUsLinkComponent
Advanced pH Meter, Sper Scientific

Advanced pH Meter, Sper Scientific

Advanced pH Meter
Catalog # 97036-026
Supplier:   Sper Scientific
Supplier #:  850055
Advanced pH Meter, Sper Scientific
Catalog # 97036-026
Supplier:  Sper Scientific
Advanced pH Meter, Sper Scientific
Advanced pH Meter, Sper Scientific

Advanced pH Meter

Price
$773.50
Log in for availability
Each
Product Details & Documents

Reads either pH (in 0.01 resolution) or mV in large digits, with time, date, and temperature (in °C or °F) shown simultaneously on the lower display.

Also indicates stable reading, low battery and displays calibration data. Scroll 99 data points with time stamps and min/max readings directly on the backlit LCD. Features hold, up to 5 point calibration, automatic buffer recognition, manual or automatic temperature compensation, and a USB port. Will accept any pH or ORP probe with a BNC connector.

Ordering information: Probe not included. Comes with 4 AAA batteries.

Specifications
  • Description:
    Advanced pH Meter
  • Cat. No.:
    97036-026
  • Dimensions:
    170 × 70 × 33 mm (6.75 × 2.75 × 1.25")
  • Weight:
    397 g (14 oz.)