Order Entry
Canada
ContactUsLinkComponent
Chloride Standard Solution, 1000 mg/L Cl- (NIST), 500 ml, Hach
Chloride Standard Solution, 1000 mg/L Cl- (NIST), 500 ml, Hach
Catalog # CA183-49
Supplier:  Hach
CAS Number:  
undefined
Chloride Standard Solution, 1000 mg/L Cl- (NIST), 500 ml, Hach
Catalog # CA183-49
Supplier:  Hach
Supplier Number:  18349
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Description:
    Chloride
  • Size:
    500 ml
  • Cat. no.:
    CA183-49
  • Supplier No.:
    183-49

Specifications

About this item

For accuracy checks in chloride determinations

Also suitable for standardizing conductivity apparatus. 1000 mg/l Cl- (1648 mg/l NaCl).