Order Entry
Canada
ContactUsLinkComponent
 
EDTA Standard Solution, 0.035 N, 100 ml, Hach
Catalog # CA2349932
Supplier:  Hach
CAS Number:  
undefined
EDTA Standard Solution, 0.035 N, 100 ml, Hach
Catalog # CA2349932
Supplier:  Hach
Supplier Number:  2349932
CAS Number:  

Specifications

  • Description:
    EDTA Standard Solution, 0.035N
  • Size:
    100 ml MDB
  • Method:
    EDTA Titration
  • Cat. no.:
    CA2349932
  • Supplier No.:
    2349932

Specifications

About this item

For hardness determination by drop-count titration.