Order Entry
Canada
ContactUsLinkComponent
Nitrogen Test Kit, Model NI-8, Hach
Nitrogen Test Kit, Model NI-8, Hach
Catalog # CA11014-416
Supplier:  Hach
CAS Number:  
undefined
Nitrogen Test Kit, Model NI-8, Hach
Catalog # CA11014-416
Supplier:  Hach
Supplier Number:  224100
CAS Number:  

Specifications

  • Chemical method:
    Nessler's reagent
  • Description:
    Nitrogen Test Kit, Model NI-8
  • Method:
    Color disc/Nessler reagent
  • Case style:
    B
  • Cat. no.:
    CA11014-416
  • Range:
    0..1 - 3.0 NH₃-N
  • Supplier No.:
    224100
  • No. of Tests:
    100

Specifications

About this item

Designed for testing ammonia in freshwater, this kit's color disc method offers low-cost, fast, reliable results in an easy-to-carry kit.

  • One of Hach's most popular test kits
  • Simple, lowcost color disc method is the most accurate visual test kit method
  • Achieve fast results with an easytocarry test kit

Contains mercury. Dispose of according to local, state and federal regulations.

Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.