ContactUsLinkComponent
Hardness (Total) Test Kit, Model 5-EP mg-L, Hach
Hardness (Total) Test Kit, Model 5-EP mg-L, Hach
Catalog # CA97026-844
Supplier:  Hach
CAS Number:  
undefined
Hardness (Total) Test Kit, Model 5-EP mg-L, Hach
Catalog # CA97026-844
Supplier:  Hach
Supplier Number:  145401
CAS Number:  
Specifications
  • Chemical method:
    EDTA
  • Description:
    Hardness (Total) Test Kit, Model 5-EP MG-L
  • Model:
    5-EP MG-L
  • Method:
    Drop count titration/ EDTA-powder pillow
  • Case style:
    B
  • Cat. no.:
    CA97026-844
  • Range:
    20 - 400 mg/l CaCO₃
  • Supplier No.:
    145401
  • No. of Tests:
    100
Specifications
About this item
Total Hardness drop count titration/EDTA test kit with range of 20-400 mg/L; contains 100 tests.
  • Utilizes drop count titration a simple, economical titration method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field