You searched for: Counting Chambers
VWR®, cell counting slides for automated cell counters
Supplier: VWR International
Slides for use with VWR® automated cell counters.
Expand 1 Items
Counting Chamber for Corning Cell Counter, 0.1 mm
Supplier: Corning
Counting Chamber for Cell Counter, reusable glass cell counting gridless slide, 0.1 mm depth same as a Neubauer Hemocytometer and comes standard with two high quality cover slip, notch for easy loading of cell suspension
Expand 1 Items
Levy Counting Chamber, Hausser Scientific
Supplier: Hausser
Hemacytometer for accurate red and white blood cell counts
Expand 1 Items
Bright-Line Counting Chamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
Expand 5 Items
Double Neubauer Counting Chamber Set, Hausser Scientific
Supplier: Hausser
Optically ground and polished milled glass chambers are diamond etched with Double improved Neubauer ruling
Expand 3 Items
Bright-Line Hemacytometer Set, Hausser Scientific
Supplier: Hausser
Double improved Neubauer counting chamber designed for accurate counting of blood cells, platelets, dust/yeast clusters, and specimens in spinal and other fluids.
Expand 4 Items
Howard Mold Counting Chamber, Hausser Scientific
Supplier: Hausser
The chamber is designed for use in determining mold counts in tomato products
Expand 1 Items
Petroff-Hausser Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
New improved Neubauer ruling pattern with 1/400 sq. mm of ruling. Designed for the counting of bacteria, sperm, blood platelets, and the content of vaccines.
Expand 5 Items
Fuchs-Resenthal Count Chamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Designed for counting leukocytes, eosinophils and cells in cerebrospinal fluid.
Expand 1 Items
Dark-Line Etched Hemacytometer, Hausser Scientific
Supplier: Hausser
The Dark-Line etched hemacytometer is designed with an H-shaped moat that forms two counting areas, or plateaus
Expand 1 Items
Petroff-Hausser Counting Chamber, Hausser Scientific
Supplier: Hausser
Designed specifically for counting bacteria, sperm, blood platelets, and the content of vaccines
Expand 2 Items
Nageotte Counting Chamber, Hausser Scientific
Supplier: Hausser
The Nageotte hemocytometer is designed to allow users to manually count low leukocyte concentrations, specifically those beyond the capabilities of a standard hemocytometer
Expand 1 Items
Counting Chambers, C-Chip, NanoEntek
Supplier: NANOENTEK USA INC.
A precision disposable hemocytometer made of sturdy quartz grade optical plastic, developed to solve the problems of conventional glass hemocytometers.
Expand 8 Items
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
With a pipetor, 15 to 20 microlilter cell-containing sample is introduced into the counting chamber through one of the ports
Expand 2 Items
Water Microscopy Counting Chamber, Sedgwick-Rafter, Hausser Scientific
Supplier: Hausser
For the microscopic examination of water or dust
Expand 2 Items
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
All complete with case and 1 pair of cover glasses.
Expand 5 Items
Counting Cells for Parasitology, Cytology and Others, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
These are used to create precise volumes of liquid to enable counting of particles in a specified volume of liquid.
Expand 2 Items
Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
Expand 3 Items
Fuchs-Rosenthal Counting Chamber, Hausser Scientific
Supplier: Hausser
Hemacytometer for counting cells in cerebrospinal fluid, etc
Expand 1 Items
Etched Double Neubauer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.
Expand 3 Items
Hemacytometer Set
Supplier: Hausser
Obtain Accurate, Repeatable Blood Cell Counts
Expand 1 Items
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
The world's first truly disposable Hemacytometer, the C-Chip offers precise volume control and is easy to use.
Expand 2 Items
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
These plastic hemacytometers offer repeatable and reliable measurements and analysis
Expand 6 Items
Miller Squares Graticle NE57, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
The Mill Sqares microscope graticle has a ratio of large to small squares of 9:1
Expand 2 Items
Adhesive Microscope Slide Grids, Diversified Biotech
Supplier: Diversified Biotech
Disposable adhesive grid stickers are ideal for counting or measuring cells and estimating the size or length of microscopic or semi-micro objects.
Expand 1 Items
Phase Hemacytometer, Hausser Scientific
Supplier: Hausser
Phase hemacytometer is used with a phase microscope for accurately counting red and white blood cells, platelets, dust/yeast clusters and specimens in spinal and other body fluids
Expand 1 Items
Adhesive Microscope Slide Wells, Diversified Biotech
Supplier: Diversified Biotech
Disposable liquid-holding wells are ideal for examining or counting tissue culture cells, yeast, bacteria, and other microrganisms.
Expand 2 Items
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
These 3×1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents.