Order Entry
Canada
Orders LinkContactUsLinkComponent
Pepstatin A

Pepstatin A

Supplier:   Bachem Americas
Pepstatin A
Supplier:  Bachem Americas
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Pepstatin A (isovaleryl-pepstatin), a low molecular weight inhibitor of acid proteases, e.g. pepsin, renin, cathepsin D, and plasmepsin II. For the Plasmodium falciparum aspartyl protease plasmepsin II, Hill et al determined an IC₅₀ of 0.1 nM.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • N-1125.0025BA
  • N-1125.0100BA
  • Protein/peptide name:
  • Pepstatin A
  • Pepstatin A
  • CAS No.:
  • 26305-03-3
  • 26305-03-3
  • MDL No.:
  • MFCD00060740
  • MFCD00060740
  • Sequence:
  • Isovaleryl-Val-Val-Sta-Ala-Sta-OH
  • Isovaleryl-Val-Val-Sta-Ala-Sta-OH
  • Molecular weight:
  • 685.90
  • 685.90
  • Molecular formula:
  • C₃₄H₆₃N₅O₉
  • C₃₄H₆₃N₅O₉
  • Storage conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $328.63
    $328.63
    Each (25mg)
       $328.63
    Minimum order quantity is 1

    Supplier Number: 4031136.0025
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    N-1125.0025BA
    Protein/peptide name
    Pepstatin A
    CAS No.
    26305-03-3
    MDL No.
    MFCD00060740
    Sequence
    Isovaleryl-Val-Val-Sta-Ala-Sta-OH
    Molecular weight
    685.90
    Molecular formula
    C₃₄H₆₃N₅O₉
    Storage conditions
    –20 ± 5 °C
  • 100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $961.65
    $961.65
    Each (100mg)
       $961.65
    Minimum order quantity is 1

    Supplier Number: 4031136.0100
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    N-1125.0100BA
    Protein/peptide name
    Pepstatin A
    CAS No.
    26305-03-3
    MDL No.
    MFCD00060740
    Sequence
    Isovaleryl-Val-Val-Sta-Ala-Sta-OH
    Molecular weight
    685.90
    Molecular formula
    C₃₄H₆₃N₅O₉
    Storage conditions
    –20 ± 5 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.