H-Phe-Leu-Glu-Glu-Leu-OH
Catalog # M-1700.0100BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:69729-06-2
- Size:100 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00076877
- Protein/peptide name:H-Phe-Leu-Glu-Glu-Leu-OH
- Molecular weight:649.74
- Sequence:H-Phe-Leu-Glu-Glu-Leu-OH
- Molecular formula:C₃₁H₄₇N₅O₁₀
- Cat. no.:M-1700.0100BA
- Supplier No.:M-1700.0100
Specifications
About this item
FLEEL, substrate of vitamin K-dependent carboxylase.