For-Met-Leu-pNA
Catalog # L-2030.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:111150-07-3
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD02259652
- Protein/peptide name:For-Met-Leu-pNA
- Molecular weight:410.49
- Sequence:For-Met-Leu-pNA
- Molecular formula:C₁₈H₂₆N₄O₅S
- Cat. no.:L-2030.0250BA
- Supplier No.:L-2030.0250
Specifications
About this item
fML-pNA, chromogenic substrate for a continuous spectrophotometric assay for peptide deformylase, see also L-2055.