Sar-Pro-Arg-pNA
Supplier: Bachem Americas
Certificates
About this item
Thrombin substrates are often cleaved as well by trypsin and trypsin-like enzymes, e.g. I-1365, I-1560, J-1120, and L-1150.
Chromogenic substrate for the assay of human α-thrombin.
Specifications
- CAS No.:77695-30-8
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD01318841
- Protein/peptide name:Sar-Pro-Arg-pNA
- Molecular weight:462.51
- Sequence:Sar-Pro-Arg-pNA
- Molecular formula:C₂₀H₃₀N₈O₅
Specifications
Frequently Bought Together