H-Lys(Abz)-Pro-Pro-pNA
50 mg
Catalog # L-1980.0050BA
Supplier: Bachem Americas
: 4027668.0050
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Internally quenched fluorogenic (FRET) substrate which allows aminopeptidase P (APP) activity determination by a continuous fluorescent assay (kcat/Km = 29'000 M⁻¹s⁻¹ for E.coli APP and kcat/Km = 1'142'000 M⁻¹s⁻¹ for rat APP).
Specifications
Specifications
- Size:50 mg
- Cat. No.:L-1980.0050BA
- Molecular formula:C₂₉H₃₇N₇O₆
- Protein/peptide name:H-Lys(Abz)-Pro-Pro-pNA
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD01318839
- Sequence:H-Lys(Abz)-Pro-Pro-pNA
- Molecular weight:579.66
- CAS No.:219138-18-8
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



