H-Val-Ala-PNA
Catalog # L-1845.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:250 mg
- Cat. no.:L-1845.0250BA
- Supplier No.:L-1845.0250
Specifications
About this item
:
:308.338 g/mol
Storage Temperature:Freezer
CAS Number:87810-63-7
Formula:C₁₄H₂₀N₄O₄
MDL Number:MFCD00058370