Order Entry
Canada
ContactUsLinkComponent
Pyr-Phe-Leu-pNA
Pyr-Phe-Leu-pNA
Supplier:  Bachem Americas
Certificates
Pyr-Phe-Leu-pNA
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    85901-57-1
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00080242
  • Protein/peptide name:
    Pyr-Phe-Leu-pNA
  • Molecular weight:
    509.56
  • Sequence:
    Pyr-Phe-Leu-pNA
  • Molecular formula:
    C₂₆H₃₁N₅O₆

Specifications

Frequently Bought Together