Z-Gly-Gly-Arg-betaNA·Hcl
Supplier: Bachem Americas
Certificates
About this item
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Z-GGR-βNA, sensitive chromogenic substrate for trypsin. Furthermore, this substrate allows a distinct histochemical localization of enteropeptidase with the trypsinogen method. It is also cleaved by neutral endopeptidase 24.5. Z-GGR-βNA is also used for determining the tryptic activity of the 20S proteasome.
Specifications
- CAS No.:1442-79-1
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038939
- Protein/peptide name:Z-Gly-Gly-Arg-betaNA · HCl
- Molecular weight:584.08
- Sequence:Z-Gly-Gly-Arg-βNA · HCl
- Molecular formula:C₂₈H₃₃N₇O₅ · HCl
Specifications
Frequently Bought Together