Order Entry
Canada
ContactUsLinkComponent
H-Ala-Phe-Pro-betaNA · HCl
H-Ala-Phe-Pro-betaNA · HCl
Supplier:  Bachem Americas
Certificates
H-Ala-Phe-Pro-betaNA · HCl
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    749831-27-4
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00237745
  • Protein/peptide name:
    H-Ala-Phe-Pro-betaNA · HCl
  • Molecular weight:
    495.02
  • Sequence:
    H-Ala-Phe-Pro-βNA · HCl
  • Molecular formula:
    C₂₇H₃₀N₄O₃ · HCl

Specifications

Frequently Bought Together