Order Entry
Canada
ContactUsLinkComponent
Boc-Leu-Ser-Thr-Arg-AMC Trifluoroacetate
Boc-Leu-Ser-Thr-Arg-AMC Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Boc-Leu-Ser-Thr-Arg-AMC Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    73554-93-5
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD09025930
  • Protein/peptide name:
    Boc-Leu-Ser-Thr-Arg-AMC Trifluoroacetate
  • Molecular weight:
    732.83
  • Sequence:
    Boc-Leu-Ser-Thr-Arg-AMC
  • Molecular formula:
    C₃₄H₅₂N₈O₁₀

Specifications

Frequently Bought Together