Fmoc-Phe-Wang resin 0.50-1.00 mmol/g 100-200 mesh
Catalog # D-2365.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. no.:D-2365.0005BA
- Supplier No.:D-2365.0005
Specifications
About this item
:
Storage Temperature:Ambient
MDL Number:MFCD00801265