Z-Ile-Glu(OtBu)-Ala-Leu-Aldehyde
Catalog # C-3900.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:158442-41-2
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00671409
- Protein/peptide name:Z-Ile-Glu(OtBu)-Ala-Leu-aldehyde
- Molecular weight:618.77
- Sequence:Z-Ile-Glu(OtBu)-Ala-Leu-aldehyde
- Molecular formula:C₃₂H₅₀N₄O₈
- Cat. no.:C-3900.0005BA
- Supplier No.:C-3900.0005
Specifications
About this item
See also Leupeptin (N-1000) and MG-132 (Calpain Inhibitor IV, N-1635).
Z-IE(OtBu)AL-CHO is an inhibitor of chymotrypsin-like activity of the multicatalytic proteinase complex (MPC; 20S proteasome) in HT4 cells. It is the first inhibitor reported so far which can cause accumulation of ubiquitinylated proteins in neuronal cells. Furthermore, this compound induced massive apoptosis in murine leukaemia L1210 cells. Therefore, proteasome inhibitors may be considered as potential anti-neoplastic agents.