Order Entry
Canada
Orders LinkContactUsLinkComponent
Boc-Phe(4-CN)-OH

Boc-Phe(4-CN)-OH

Boc-Phe(4-CN)-OH
Supplier:  
Certificates
Boc-Phe(4-CN)-OH

0 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: Boc-4-cyano-Phe-OH

Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Derivative for introducing p-cyanophenylalanine during Boc-SPPS. This Phe derivative may serve as a selectively excitable fluorophore or as an IR probe in peptides and for obtaining FRET substrates.

Synonyms:  Boc-4-cyano-Phe-OH, N-Boc-4-cyano-L-phenylalanine, Boc-Phe(4-CN)-OH, N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine
  • CAS Number:131724-45-3
  • Formula:C₁₅H₁₈N₂O₄
  • MDL Number:MFCD00672526
  • Molecular Weight:290.319 g/mol
  • Storage Temperature:Freezer
Product Family Options