Nitrogen-Nitrate Reagent Set, LR, 10 ml, Cadmium Reduction, Hach
Catalog # CA24298-00
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Cadmium reduction
- Description:Nitrogen-Nitrate Reagent Set, LR, 10 mL, Cadmium Reduction
- Method:8192 - Cadmium Reduction
- Instrument:All except DR/3000, DR/2010, DR/2000, PC II
- Cat. no.:CA24298-00
- Range:0.01 - 0.50 mg/l NO₃-N
- Supplier No.:24298-00
- No. of Tests:100
Specifications
About this item
For determination of Nitrate Nitrogen by the Cadmium Reduction method
Hach Method 8192. Range: 0.01 - 0.50 mg/L NO3-N. Sample Size: 10 mL.. Set contains NitraVer® 6 and NitriVer® 3 Nitrite Reagent Powder Pillows for 100 tests.