Order Entry
Canada
ContactUsLinkComponent
Alkali Solution for Calcium and Magnesium, 100 ml, Hach
Alkali Solution for Calcium and Magnesium, 100 ml, Hach
Catalog # CA22417-32
Supplier:  Hach
CAS Number:  
undefined
Alkali Solution for Calcium and Magnesium, 100 ml, Hach
Catalog # CA22417-32
Supplier:  Hach
Supplier Number:  2241732
CAS Number:  

Specifications

  • Chemical method:
    Calmagite
  • Description:
    Alkali Solution for Calcium and Magnesium
  • Size:
    100 mL Marked Dropping Bottle
  • Method:
    8030 - Calmagite Colorimetric
  • Cat. no.:
    CA22417-32
  • Supplier No.:
    22417-32

Specifications

About this item

For determination of calcium and magnesium hardness in water by the Calmagite Colorimetric method

Hach Method 8030.

Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.