Order Entry
Canada
ContactUsLinkComponent
Phosphorus, Orthophosphate (reactive) Test Kit, Model PO-19, Hach
Phosphorus, Orthophosphate (reactive) Test Kit, Model PO-19, Hach
Catalog # CA2248-00
Supplier:  Hach
CAS Number:  
undefined
Phosphorus, Orthophosphate (reactive) Test Kit, Model PO-19, Hach
Catalog # CA2248-00
Supplier:  Hach
Supplier Number:  224800
CAS Number:  

Specifications

  • Chemical method:
    Ascorbic acid
  • Description:
    Phosphorus, Orthophosphate (reactive) Test Kit, Model PO-19
  • Model:
    PO-19
  • Method:
    Color disc/Ascorbic acid
  • Case style:
    R
  • Increments:
    Smallest steps - 0.02
  • Cat. no.:
    CA2248-00
  • Range:
    0 - 4 mg/l PO₄, 0 - 40 mg/l PO₄, 0 - 0.8 mg/l PO₄
  • Supplier No.:
    2248-00
  • No. of Tests:
    100

Specifications

About this item

Method / Chemistry: Color disc / Ascorbic acid.

  • Results in less than 5 minutes for clean water samples
  • Utilizes simple color disc comparator for accurate measurements
  • Model PO19A contains filtration apparatus for turbid samples

Range (mg/L): 0-1, Smallest Increment- 0.02. Range (mg/L): 0-5, Smallest Increment- 0.1. Range (mg/L): 0-40, Smallest Increment- 1. Approx. Number of Tests: 100. Case Style: 12x7.25x6 inches Blue Polypropylene Carrying Case.