BOD Standard Solution, 3000 mg/L, 10-ml Voluette« Ampules, Hach
CA11017-030
: Hach
:
- Concentration:3000 mg/L
- Pack type:Voluette Ampules
- Description:BOD Standard Solution
- Size:10 ml
- Cat. no.:CA11017-030
- Supplier No.:1486610
For accuracy checks in Biochemical Oxygen Demand (BOD) determinations
- Simple Accuracy Checks
- Minimize waste
- Reliable results
3000 mg/L Glucose plus Glutamic Acid.
: Pack of 16 10-mL ampules.