Phenol Reagent Powder Pillows (for nonextraction method), Hach
Catalog # CA89231-096
Supplier: Hach
CAS Number:
Specifications
- Chemical method:4AAP (4-Aminoantipyrine)
- Description:Phenol Reagent Powder Pillows (for nonextraction method)
- Method:4-Aminoantipyrine
- Cat. no.:CA89231-096
- Supplier No.:2481569
Specifications
About this item
For determination of phenols by the 4-Aminoantipyrine Method.
Ordering information: 100/pk.