Mehlich 2 Soil Extractant Solution, 500 ml, Hach
Catalog # CA89231-024
Supplier: Hach
CAS Number:
Specifications
- Description:Mehlich 2 Soil Extractant Solution
- Size:500 mL
- Cat. no.:CA89231-024
- Supplier No.:2266349
Specifications
About this item
Extractant for phosphate and potassium determination in soils.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.