Nitrogen-Nitrate Reagent Set, LR, 25 ml, Cadmium Reduction, Hach
Catalog # CA22451-00
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Cadmium reduction
- Description:Nitrogen-Nitrate Reagent Set, LR, 25 mL, Cadmium Reduction
- Method:8192 - Cadmium Reduction
- Cat. no.:CA22451-00
- Range:0 - 0.50 mg/l NO₃--N
- Supplier No.:22451-00
- No. of Tests:100
Specifications
About this item
For determination of Nitrate Nitrogen by the Cadmium Reduction method
Hach Method 8192. Range: up to 0.50 mg/L NO3-N. Sample Size: 25 mL. Reagent Set includes: NitriVer® 3 Reagent Powder Pillows and NitraVer® 6 Reagent Powder Pillows for 100 tests.