ContactUsLinkComponent
Hardness (Total) Reagent Set for HA-71A Test Kit, Hach
Hardness (Total) Reagent Set for HA-71A Test Kit, Hach
Catalog # CA89231-078
Supplier:  Hach
CAS Number:  
undefined
Hardness (Total) Reagent Set for HA-71A Test Kit, Hach
Catalog # CA89231-078
Supplier:  Hach
Supplier Number:  2437800
CAS Number:  
Specifications
  • Chemical method:
    EDTA
  • Description:
    Hardness (Total) Reagent Set for HA-71A Test Kit
  • Size:
    1 Kit
  • Cat. no.:
    CA89231-078
  • Supplier No.:
    2437800
  • No. of Tests:
    100
Specifications
About this item
For determination of Total Hardness by the Drop Count EDTA Titration Method
Replacement reagent set for HA-71A Test Kit. Set contains Buffer Solution (Hardness 1), Hardness 2 and Hardness 3 Test Solutions for approximately 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.