Order Entry
Canada
ContactUsLinkComponent
pPb-4, Neutralizer, 22 ml, Hach
pPb-4, Neutralizer, 22 ml, Hach
Catalog # CA2368855
Supplier:  Hach
CAS Number:  
undefined
pPb-4, Neutralizer, 22 ml, Hach
Catalog # CA2368855
Supplier:  Hach
Supplier Number:  2368855
CAS Number:  

Specifications

  • Chemical method:
    LeadTrak
  • Description:
    pPb-4, Neutralizer
  • Size:
    22 mL
  • Method:
    LeadTrak
  • Cat. no.:
    CA2368855
  • Supplier No.:
    2368855
  • No. of Tests:
    20

Specifications

About this item

For Lead determination by the LeadTrak™ Fast Column Extraction method

Hach method 8317.

Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.