Specifications
- Max. speed:100–3000 rpm
- Description:Low Speed Benchtop Centrifuge LMC-3000
- Speed increment:100 rpm
- Electrical:120 V, 60 Hz
- Max. RCF:1700 x g
- Dimensions:235 x 420 x 495 mm
- Operating temperature:4...65 °C (39.2...149 °F)
- Timer:0–90 min.
- Weight:13.5 kg (28.6 lbs.)
- Cat. no.:89194-510
- Supplier No.:LMC-3000 USA
Specifications
About this item
The Grant Bio LMC-3000 is a low speed benchtop centrifuge designed for biomedical applications
Spin speed: up to 3000rpm for tubes, up to 2000rpm for microtitre plates
Timed operation (1 to 90 minutes), with automatic switch-off
‘Soft-start’ and ‘run-down’ of the rotor
Choice of interchangeable rotors for up to 12 centrifuge tubes or 2 microplates
Auto ‘IMBALANCE’ stop and warning diagnostics
Low noise level
Sturdy metal housing and lid, automatic imbalance switch-off and lid lock when centrifuge is running providing safe operation in your workplace
The centrifuge is suitable for a wide range of analytical applications including biomedical, bio-organic, and immunoenzyme analysis.
It features interchangeable rotors for accommodating centrifuge tubes (10 to 15mL, or 50mL) or standard 96-well microtitre plates.