ContactUsLinkComponent
Distillation Receiver, Use At Other Than Atmospheric Pressure, Ace Glass
Distillation Receiver, Use At Other Than Atmospheric Pressure, Ace Glass
Catalog # 89074-456
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Distillation Receiver, Use At Other Than Atmospheric Pressure, Ace Glass
Catalog # 89074-456
Supplier:  Ace Glass
Supplier Number:  9375-08
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Distillation Receiver
  • Cat. no.:
    89074-456
  • Supplier No.:
    9375-08
Specifications
About this item
This receiver is designed for operation at other than atmospheric pressure
Vacuum is applied to the side arm with the T-bore stopcock. The secondary receiver is vented by means of the lower stopcock. The capacity from the bottom of the drip to the stopcock is 2.5 mL. The apparatus has a [ST] 14/20 outer joint at top, [ST] 14/20 inner joint at bottom. Hose connections are for use with 3/8-inch I.D. Both stopcocks are glass. Fabricated from borosilicate glass.