Hardness (Calcium) Reagent Set, Buret, Hach
CA24470-00
: Hach
:
- Chemical method:EDTA
- Description:Hardness (Calcium) Reagent Set, Buret
- Method:8222 - EDTA Titration
- Compliance:EPA - Yes
- Cat. no.:CA24470-00
- Range:0 - 5000 mg/l CaCO₃
- Supplier No.:2447000
- No. of Tests:100
Reagent set for the determination of Calcium Hardness by EDTA Buret Titration Method (Hach method 8222)
EPA Accepted method. Range: 0-5,000 mg/L as CaCO3. Set contains CalVer® 2 Calcium Indicator Powder Pillows, Potassium Hydroxide Solution (8 N) and TitraVer® (EDTA) Titrant for up to approximately 100 tests.
: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.