Order Entry
Canada
ContactUsLinkComponent
Molybdate Test Kit, Model MO-LR, Hach
Molybdate Test Kit, Model MO-LR, Hach
Catalog # CA23593-00
Supplier:  Hach
CAS Number:  
undefined
Molybdate Test Kit, Model MO-LR, Hach
Catalog # CA23593-00
Supplier:  Hach
Supplier Number:  2359300
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Ternary complex
  • Description:
    Molybdate Test Kit, Model MO-LR
  • Model:
    MO-LR
  • Method:
    Color disc/Ternary complex
  • Case style:
    D
  • Cat. no.:
    CA23593-00
  • Range:
    0.25 - 3.0 mg/l Mo
  • Supplier No.:
    23593-00
  • No. of Tests:
    100

Specifications

About this item

Molybdates are gaining over chromates for use as corrosion inhibitors due to lower toxicity.

  • Kit reagents react with Molybdate to form a yellow color, which is measured with an easytouse color disc method
  • Effectively monitors corrosion inhibitors

The high cost of these treatments warrant careful control of their concentration. Kit reagents react with Molybdate to form a yellow color, which is measured with a color disc.