Iron Color Disc Test Kit, Model IR-18B, Hach
Catalog # CA97026-884
Supplier: Hach
CAS Number:
Specifications
- Chemical method:1,10-Phenanthroline
- Description:Iron Color Disc Test Kit, Model IR-18B
- Model:IR-18B
- Method:Color disc/ 1,10 Phenanthroline
- Case style:B
- Cat. no.:CA97026-884
- Range:0.2 - 7 mg/l Fe
- Supplier No.:146401
- No. of Tests:100
Specifications
About this item
Using the simple color disc method, this kit measures Iron (medium range - 0.2-10 mg/L) and contains 100 tests.
- Simple, lowcost color disc method is the most accurate visual test kit method
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field