Order Entry
Canada
ContactUsLinkComponent
Corning® Mini Bioreactor Centrifuge Tube, Corning
Corning® Mini Bioreactor Centrifuge Tube, Corning
Catalog # 30618-034
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
Corning® Mini Bioreactor Centrifuge Tube, Corning
Catalog # 30618-034
Supplier:  Corning
Supplier Number:  431720
Product available on GSA Advantage®
   Low Manufacturing Impact

Specifications

  • Cap Type:
    Vented cap
  • Sterility:
    Sterile
  • Volume:
    50 ml (1.7 oz.)
  • Description:
    Mini Bioreactor Centrifuge Tube
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    30618-034
  • Packaging:
    25/Rack
  • Supplier No.:
    431720

Specifications

About this item

PP, translucent, with orange PP screw cap.

  • Large marking spot enables clear identification and experimental parameters
  • Easy-to-read graduations
  • Non pyrogenic
  • Free from RNase and DNase
  • Sterile

Ideal for high throughput process optimization for suspension cell culture, the Mini Bioreactor consists of a centrifuge tube with vented cap. Tube applications include cell line developement, clone selection, media optimization, and recombinant protein development. Screw cap features four vent holes and a 0.22 µm hydrophobic membrane to allow for gas exchange.