Order Entry
Canada
ContactUsLinkComponent
DEHA Reagent 2 Solution, 100 ml, Hach
DEHA Reagent 2 Solution, 100 ml, Hach
  CA97026-596
 :  Hach
 :  
DEHA Reagent 2 Solution, 100 ml, Hach
  CA97026-596
 :  Hach
 :  2168042
 :  

 

  • Chemical method:
    Iron reduction
  • Description:
    DEHA Reagent 2 Solution
  • Size:
    100 mL
  • Method:
    8140 - Iron Reduction
  • Cat. no.:
    CA97026-596
  • No. of Tests:
    100

 

 

For determination of oxygen scavengers by the iron reduction method

Hach method 8140 determines carbohydrazide, diethylhydroxylamine (DEHA), hydroquinone, iso-ascorbic acid (erythorbic acid) and methylethylketoxime.

 : Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.