Orders LinkContactUsLinkComponent
LeadTrak™ Reagent Set, Fast Column Extraction Method, Hach
LeadTrak™ Reagent Set, Fast Column Extraction Method, Hach
LeadTrak™ Reagent Set, Fast Column Extraction Method, 20 tests, 100 mL, LeadTrak, 5 - 150 μg/l Pb, 20
Catalog # CA23750-00
Supplier:   Hach
Supplier #:  2375000
LeadTrak™ Reagent Set, Fast Column Extraction Method, Hach
Catalog # CA23750-00
Supplier:  Hach
CAS Number:  
LeadTrak™ Reagent Set, Fast Column Extraction Method, Hach
LeadTrak™ Reagent Set, Fast Column Extraction Method, Hach
LeadTrak™ Reagent Set, Fast Column Extraction Method, 20 tests, 100 mL, LeadTrak, 5 - 150 μg/l Pb, 20
CAS Number:  
Price
$814.81
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Reagent Set for Lead determination by the LeadTrak™ Fast Column Extraction
Hach Method 8317. Range: 5 to 150 µg/L as Pb. Sample size: 100 mL.
Ordering information: 20 tests per reagent set. Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Documents
Specifications
Specifications
  • Chemical method:
    LeadTrak
  • Description:
    LeadTrak™ Reagent Set, Fast Column Extraction Method, 20 tests
  • Size:
    100 mL
  • Range:
    5 - 150 μg/l Pb
  • No. of tests:
    20
  • Cat. No.:
    CA23750-00
  • Instrument:
    All except DR/800
  • Method:
    8317 - LeadTrak
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)