pH Test Kit, 5.5 - 8.5 pH, Model 17F, Hach
Catalog # CA11024-464
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Bromthymol blue
- Description:pH Test Kit, 5.5 - 8.5 pH, Model 17F
- Model:17F
- Method:Color disc/Bromthymol blue
- Case style:B
- Cat. no.:CA11024-464
- Range:pH 5.6 - 8.4
- Supplier No.:147006
- No. of Tests:100
Specifications
About this item
-Simple, low-cost color disc method is the most accurate visual test kit method
-Achieve fast results with an easy-to-carry test kit
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.