ContactUsLinkComponent
Hardness (Total) Test Kit, Model HA-DT, Hach
Hardness (Total) Test Kit, Model HA-DT, Hach
  CA11014-504
 :  Hach
 :  
undefined
Hardness (Total) Test Kit, Model HA-DT, Hach
  CA11014-504
 :  Hach
 :  2063600
 :  
  • Chemical method:
    EDTA
  • Description:
    Hardness (Total) Test Kit, Model HA-DT
  • Model:
    HA-DT
  • Method:
    Digital Titrator/EDTA
  • Case style:
    F
  • Increments:
    smallest steps - 0.1-10
  • Cat. no.:
    CA11014-504
  • Range:
    10 - 4000 mg/l CaCO₃
  • Supplier No.:
    2063600
  • No. of Tests:
    100
Utilizing the Hach Digital Titrator, the Model HA-DT kit is used to determine hardness concentrations.
  • Hach Digital Titrator offers higher levels of precision and accuracy than drop count procedures, with reproducibility comparable to a buret titration
  • Rugged titration device is ideal for field use, with no glass burets required
Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10-4,000 mg/L. Contains approximately 100 tests.