SINGLET™ Single-Use pH Buffers, Hach
About this item
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
- Ideal for on-site analysis
- Easy to use
- Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
Ordering information: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
5 Options Available Below
Product Details & Documents
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
- Ideal for on-site analysis
- Easy to use
- Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
Ordering information: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA11014-252
- CA97021-424
- CA11014-256
- CA11014-254
- CA11028-036
- Concentration:
- 4.01 pH
- 7.00 pH
- 10.01 pH
Specifications
Product Family Options
Product Information
- pH accuracypHAvailabilityPrice
- Supplier Number: 2770020Specifications:
More
Specifications
Cat. No.CA11014-252Concentration4.01 pH - Supplier Number: 2770120Specifications:
More
Specifications
Cat. No.CA97021-424Concentration7.00 pH - Supplier Number: 2770220Specifications:
More
Specifications
Cat. No.CA11014-256Concentration10.01 pH - CA11014-254Combination PackSupplier Number: 2769920Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.CA11014-254Concentration - CA11028-036Combination PackSupplier Number: 2769820Specifications:
More
Specifications
Cat. No.CA11028-036Concentration
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



